Methyl-2-[4-(3-piperidinylmethoxy)phenyl]acetate
Supplier: Matrix Scientific
023025-500MG
101815-200EA
315.38
USD
101815-200
Methyl-2-[4-(3-piperidinylmethoxy)phenyl]acetate
Methyl-2-[4-(3-piperidinylmethoxy)phenyl]acetate
Hazard: Irritant
Formula:
C₁₅H₂₁NO₃ |
MDL Number:
MFCD08688362 CAS Number: 946787-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...