3-{[4-(1-Methyl-1-phenylethyl)phenoxy]methyl}piperidine

Supplier: Matrix Scientific

023020-500MG
101815-190EA 235.75 USD
101815-190
3-{[4-(1-Methyl-1-phenylethyl)phenoxy]methyl}piperidine
3-{[4-(1-Methyl-1-phenylethyl)phenoxy]methyl}piperidine

Hazard: Irritant

Formula: C₂₁H₂₇NO
MDL Number: MFCD08688341
CAS Number: 946787-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR