(4-Chlorophenyl)[4-(3-piperidinylmethoxy)phenyl]methanone

Supplier: Matrix Scientific

023019-500MG
101815-188EA 235.75 USD
101815-188
(4-Chlorophenyl)[4-(3-piperidinylmethoxy)phenyl]methanone
(4-Chlorophenyl)[4-(3-piperidinylmethoxy)phenyl]methanone

Hazard: Irritant

Formula: C₁₉H₂₀ClNO₂
MDL Number: MFCD08687458
CAS Number: 946724-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR