3-[(4-Ethoxyphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023018-500MG
101815-186EA 315.38 USD
101815-186
3-[(4-Ethoxyphenoxy)methyl]piperidine
3-[(4-Ethoxyphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₄H₂₁NO₂
MDL Number: MFCD08687737
CAS Number: 946713-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR