3-[(4-Chloro-3,5-dimethylphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023015-500MG
101815-180EA 315.38 USD
101815-180
3-[(4-Chloro-3,5-dimethylphenoxy)methyl]piperidine
3-[(4-Chloro-3,5-dimethylphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₄H₂₀ClNO
MDL Number: MFCD08687282
CAS Number: 946787-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR