3-[(4-Fluorophenoxy)methyl]piperidine

Supplier: Matrix Scientific

023013-500MG
101815-176EA 315.38 USD
101815-176
3-[(4-Fluorophenoxy)methyl]piperidine
3-[(4-Fluorophenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₂H₁₆FNO
MDL Number: MFCD06247688
CAS Number: 142220-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR