3-[(3-Ethylphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023011-500MG
101815-172EA 315.38 USD
101815-172
3-[(3-Ethylphenoxy)methyl]piperidine
3-[(3-Ethylphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD08687778
CAS Number: 946713-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR