3-[(3-Methylphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023008-500MG
101815-166EA 235.75 USD
101815-166
3-[(3-Methylphenoxy)methyl]piperidine
3-[(3-Methylphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD06248327
CAS Number: 693764-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR