3-[(3-Isopropylphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023007-500MG
101815-164EA 315.38 USD
101815-164
3-[(3-Isopropylphenoxy)methyl]piperidine
3-[(3-Isopropylphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₅H₂₃NO
MDL Number: MFCD08687997
CAS Number: 946724-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR