3-[(3-Iodophenoxy)methyl]piperidine

Supplier: Matrix Scientific

023003-500MG
101815-156EA 315.38 USD
101815-156
3-[(3-Iodophenoxy)methyl]piperidine
3-[(3-Iodophenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₂H₁₆INO
MDL Number: MFCD08687940
CAS Number: 946787-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR