3-[(2,5-Dimethylphenoxy)methyl]piperidine
Supplier: Matrix Scientific
022996-500MG
101815-142EA
315.38
USD
101815-142
3-[(2,5-Dimethylphenoxy)methyl]piperidine
3-[(2,5-Dimethylphenoxy)methyl]piperidine
Hazard: Irritant
Formula:
C₁₄H₂₁NO |
MDL Number:
MFCD08687650 CAS Number: 946787-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...