2-(2-Benzylphenoxy)-5-chloroaniline
Supplier: Matrix Scientific
021982-500MG
101813-126EA
315.38
USD
101813-126
2-(2-Benzylphenoxy)-5-chloroaniline
2-(2-Benzylphenoxy)-5-chloroaniline
Hazard: Irritant
Formula:
C₁₉H₁₆ClNO |
MDL Number:
MFCD08686889 CAS Number: 946772-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...