5-Chloro-2-(2,5-dimethylphenoxy)aniline

Supplier: Matrix Scientific

021981-500MG
101813-124EA 315.38 USD
101813-124
5-Chloro-2-(2,5-dimethylphenoxy)aniline
5-Chloro-2-(2,5-dimethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄ClNO
MDL Number: MFCD07365155
CAS Number: 893751-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR