5-Chloro-2-(2,4-dimethylphenoxy)aniline

Supplier: Matrix Scientific

021979-500MG
101813-120EA 315.38 USD
101813-120
5-Chloro-2-(2,4-dimethylphenoxy)aniline
5-Chloro-2-(2,4-dimethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄ClNO
MDL Number: MFCD08687275
CAS Number: 937606-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR