5-Chloro-2-(2,3-dimethylphenoxy)aniline

Supplier: Matrix Scientific

021978-500MG
101813-118EA 235.75 USD
101813-118
5-Chloro-2-(2,3-dimethylphenoxy)aniline
5-Chloro-2-(2,3-dimethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄ClNO
MDL Number: MFCD07365152
CAS Number: 893750-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR