5-Chloro-2-(2-methylphenoxy)aniline

Supplier: Matrix Scientific

021977-500MG
101813-116EA 235.75 USD
101813-116
5-Chloro-2-(2-methylphenoxy)aniline
5-Chloro-2-(2-methylphenoxy)aniline

Hazard: Irritant

Formula: C₁₃H₁₂ClNO
MDL Number: MFCD07365156
CAS Number: 3169-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR