2-([1,1'-Biphenyl]-2-yloxy)-5-chloroaniline

Supplier: Matrix Scientific

021975-500MG
101813-112EA 315.38 USD
101813-112
2-([1,1'-Biphenyl]-2-yloxy)-5-chloroaniline
2-([1,1'-Biphenyl]-2-yloxy)-5-chloroaniline

Hazard: Irritant

Formula: C₁₈H₁₄ClNO
MDL Number: MFCD08686931
CAS Number: 946727-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR