N-[2-(2-Amino-4-chlorophenoxy)ethyl]-N-methyl-N-phenylamine

Supplier: Matrix Scientific

021966-500MG
101813-094EA 315.38 USD
101813-094
N-[2-(2-Amino-4-chlorophenoxy)ethyl]-N-methyl-N-phenylamine
N-[2-(2-Amino-4-chlorophenoxy)ethyl]-N-methyl-N-phenylamine

Hazard: Irritant

Formula: C₁₅H₁₇ClN₂O
MDL Number: MFCD08686781
CAS Number: 946715-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR