5-Chloro-2-(tetrahydro-2H-pyran-2-ylmethoxy)phenylamine

Supplier: Matrix Scientific

021965-500MG
101813-092EA 315.38 USD
101813-092
5-Chloro-2-(tetrahydro-2H-pyran-2-ylmethoxy)phenylamine
5-Chloro-2-(tetrahydro-2H-pyran-2-ylmethoxy)phenylamine

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₂
MW: 241.72 g/mol
MDL Number: MFCD08687481
CAS Number: 946727-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR