5-Chloro-2-[2-(1-piperidinyl)ethoxy]phenylamine

Supplier: Matrix Scientific

021964-500MG
101813-090EA 315.38 USD
101813-090
5-Chloro-2-[2-(1-piperidinyl)ethoxy]phenylamine
5-Chloro-2-[2-(1-piperidinyl)ethoxy]phenylamine

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD07365168
CAS Number: 893751-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR