5-Chloro-2-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

021962-500MG
101813-086EA 315.38 USD
101813-086
5-Chloro-2-(tetrahydro-2-furanylmethoxy)aniline
5-Chloro-2-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₂
MDL Number: MFCD07365161
CAS Number: 893751-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR