5-Chloro-2-[(2-methylpentyl)oxy]aniline

Supplier: Matrix Scientific

021961-500MG
101813-084EA 315.38 USD
101813-084
5-Chloro-2-[(2-methylpentyl)oxy]aniline
5-Chloro-2-[(2-methylpentyl)oxy]aniline

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD08687374
CAS Number: 946772-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR