5-Chloro-2-(1-naphthylmethoxy)aniline
Supplier: Matrix Scientific
021958-500MG
101813-078EA
315.38
USD
101813-078
5-Chloro-2-(1-naphthylmethoxy)aniline
5-Chloro-2-(1-naphthylmethoxy)aniline
Hazard: Irritant
Formula:
C₁₇H₁₄ClNO |
MDL Number:
MFCD08687402 CAS Number: 946682-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...