5-Chloro-2-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

021955-500MG
101813-072EA 315.38 USD
101813-072
5-Chloro-2-(3-phenylpropoxy)aniline
5-Chloro-2-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₁₅H₁₆ClNO
MDL Number: MFCD08687464
CAS Number: 946714-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR