5-Chloro-2-(phenethyloxy)aniline

Supplier: Matrix Scientific

021951-500MG
101813-064EA 315.38 USD
101813-064
5-Chloro-2-(phenethyloxy)aniline
5-Chloro-2-(phenethyloxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄ClNO
MDL Number: MFCD07365166
CAS Number: 648926-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR