5-Chloro-2-[2-(2-methoxyethoxy)ethoxy]aniline

Supplier: Matrix Scientific

021950-500MG
101813-062EA 235.75 USD
101813-062
5-Chloro-2-[2-(2-methoxyethoxy)ethoxy]aniline
5-Chloro-2-[2-(2-methoxyethoxy)ethoxy]aniline

Hazard: Irritant

Formula: C₁₁H₁₆ClNO₃
MDL Number: MFCD08687359
CAS Number: 946727-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR