5-Chloro-2-[2-(diethylamino)ethoxy]aniline

Supplier: Matrix Scientific

021948-500MG
101813-058EA 235.75 USD
101813-058
5-Chloro-2-[2-(diethylamino)ethoxy]aniline
5-Chloro-2-[2-(diethylamino)ethoxy]aniline

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂O
MDL Number: MFCD08687257
CAS Number: 96079-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR