N-[2-(2-Amino-4-chlorophenoxy)ethyl]-N,N-dimethylamine

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-(2-(dimethylamino)ethoxy)aniline

021947-500MG
101813-056EA 315.38 USD
101813-056
N-[2-(2-Amino-4-chlorophenoxy)ethyl]-N,N-dimethylamine
N-[2-(2-Amino-4-chlorophenoxy)ethyl]-N,N-dimethylamine

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MW: 214.7 g/mol
MDL Number: MFCD08686778
CAS Number: 631862-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR