5-Chloro-2-(cyclohexylmethoxy)aniline

Supplier: Matrix Scientific

021945-500MG
101813-052EA 315.38 USD
101813-052
5-Chloro-2-(cyclohexylmethoxy)aniline
5-Chloro-2-(cyclohexylmethoxy)aniline

Hazard: Irritant

Formula: C₁₃H₁₈ClNO
MDL Number: MFCD08687247
CAS Number: 946714-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR