3-Chloro-2-(2-naphthyloxy)aniline

Supplier: Matrix Scientific

021940-500MG
101813-042EA 235.75 USD
101813-042
3-Chloro-2-(2-naphthyloxy)aniline
3-Chloro-2-(2-naphthyloxy)aniline

Hazard: Irritant

Formula: C₁₆H₁₂ClNO
MDL Number: MFCD08687405
CAS Number: 937604-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR