3-Chloro-2-(2,3-dihydro-1H-inden-5-yloxy)aniline

Supplier: Matrix Scientific

021937-500MG
101813-036EA 315.38 USD
101813-036
3-Chloro-2-(2,3-dihydro-1H-inden-5-yloxy)aniline
3-Chloro-2-(2,3-dihydro-1H-inden-5-yloxy)aniline

Hazard: Irritant

Formula: C₁₅H₁₄ClNO
MDL Number: MFCD08687264
CAS Number: 937604-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR