Methyl-2-[4-(2-amino-6-chlorophenoxy)phenyl]acetate

Supplier: Matrix Scientific

021935-500MG
101813-032EA 235.75 USD
101813-032
Methyl-2-[4-(2-amino-6-chlorophenoxy)phenyl]acetate
Methyl-2-[4-(2-amino-6-chlorophenoxy)phenyl]acetate

Hazard: Irritant

Formula: C₁₅H₁₄ClNO₃
MDL Number: MFCD08688126
CAS Number: 946714-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR