2-[4-(sec-Butyl)phenoxy]-3-chloroaniline

Supplier: Matrix Scientific

021931-500MG
101813-024EA 235.75 USD
101813-024
2-[4-(sec-Butyl)phenoxy]-3-chloroaniline
2-[4-(sec-Butyl)phenoxy]-3-chloroaniline

Hazard: Irritant

Formula: C₁₆H₁₈ClNO
MDL Number: MFCD08687172
CAS Number: 946772-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR