3-Chloro-2-(4-ethoxyphenoxy)aniline

Supplier: Matrix Scientific

021925-500MG
101813-012EA 315.38 USD
101813-012
3-Chloro-2-(4-ethoxyphenoxy)aniline
3-Chloro-2-(4-ethoxyphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄ClNO₂
MDL Number: MFCD08687295
CAS Number: 937604-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR