3-Chloro-2-(4-methoxyphenoxy)phenylamine

Supplier: Matrix Scientific

021924-500MG
101813-010EA 224.25 USD
101813-010
3-Chloro-2-(4-methoxyphenoxy)phenylamine
3-Chloro-2-(4-methoxyphenoxy)phenylamine

Hazard: Irritant

Formula: C₁₃H₁₂ClNO₂
MDL Number: MFCD08687365
CAS Number: 937604-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR