3-Chloro-2-(4-fluorophenoxy)aniline

Supplier: Matrix Scientific

021920-500MG
101813-002EA 235.75 USD
101813-002
3-Chloro-2-(4-fluorophenoxy)aniline
3-Chloro-2-(4-fluorophenoxy)aniline

Hazard: Irritant

Formula: C₁₂H₉ClFNO
MDL Number: MFCD08687310
CAS Number: 937604-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR