3-Chloro-2-(3-methylphenoxy)aniline

Supplier: Matrix Scientific

021916-500MG
101812-994EA 235.75 USD
101812-994
3-Chloro-2-(3-methylphenoxy)aniline
3-Chloro-2-(3-methylphenoxy)aniline

Hazard: Irritant

Formula: C₁₃H₁₂ClNO
MDL Number: MFCD08687377
CAS Number: 937604-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR