Methyl-3-(2-amino-6-chlorophenoxy)benzoate

Supplier: Matrix Scientific

021913-500MG
101812-988EA 235.75 USD
101812-988
Methyl-3-(2-amino-6-chlorophenoxy)benzoate
Methyl-3-(2-amino-6-chlorophenoxy)benzoate

Hazard: Irritant

Formula: C₁₄H₁₂ClNO₃
MDL Number: MFCD08688120
CAS Number: 946682-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR