3-Chloro-2-(3-chlorophenoxy)aniline
Supplier: Matrix Scientific
021908-500MG
101812-978EA
315.38
USD
101812-978
3-Chloro-2-(3-chlorophenoxy)aniline
3-Chloro-2-(3-chlorophenoxy)aniline
Hazard: Irritant
Formula:
C₁₂H₉Cl₂NO |
MDL Number:
MFCD08687240 CAS Number: 946772-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...