2-(2-Benzylphenoxy)-3-chloroaniline

Supplier: Matrix Scientific

021905-500MG
101812-972EA 235.75 USD
101812-972
2-(2-Benzylphenoxy)-3-chloroaniline
2-(2-Benzylphenoxy)-3-chloroaniline

Hazard: Irritant

Formula: C₁₉H₁₆ClNO
MDL Number: MFCD08686888
CAS Number: 946727-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR