3-Chloro-2-(2,4-dimethylphenoxy)aniline

Supplier: Matrix Scientific

021902-500MG
101812-966EA 235.75 USD
101812-966
3-Chloro-2-(2,4-dimethylphenoxy)aniline
3-Chloro-2-(2,4-dimethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄ClNO
MDL Number: MFCD08687270
CAS Number: 937604-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR