2-([1,1'-Biphenyl]-2-yloxy)-3-chloroaniline
Supplier: Matrix Scientific
021898-500MG
101812-958EA
315.38
USD
101812-958
2-([1,1'-Biphenyl]-2-yloxy)-3-chloroaniline
2-([1,1'-Biphenyl]-2-yloxy)-3-chloroaniline
Hazard: Irritant
Formula:
C₁₈H₁₄ClNO |
MDL Number:
MFCD08686930 CAS Number: 946772-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...