3-Chloro-2-(2-ethoxyphenoxy)aniline

Supplier: Matrix Scientific

021897-500MG
101812-956EA 315.38 USD
101812-956
3-Chloro-2-(2-ethoxyphenoxy)aniline
3-Chloro-2-(2-ethoxyphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄ClNO₂
MDL Number: MFCD08687293
CAS Number: 946682-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR