3-Chloro-2-(2-chlorophenoxy)aniline

Supplier: Matrix Scientific

021894-500MG
101812-950EA 315.38 USD
101812-950
3-Chloro-2-(2-chlorophenoxy)aniline
3-Chloro-2-(2-chlorophenoxy)aniline

Hazard: Irritant

Formula: C₁₂H₉Cl₂NO
MDL Number: MFCD08687239
CAS Number: 946772-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR