3-Chloro-2-phenoxyaniline

Supplier: Matrix Scientific

021892-500MG
101812-946EA 402.5 USD
101812-946
3-Chloro-2-phenoxyaniline
3-Chloro-2-phenoxyaniline

Hazard: Irritant

Formula: C₁₂H₁₀ClNO
MDL Number: MFCD08687423
CAS Number: 3169-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR