3-Chloro-2-(2-propoxyethoxy)aniline

Supplier: Matrix Scientific

021891-500MG
101812-944EA 315.38 USD
101812-944
3-Chloro-2-(2-propoxyethoxy)aniline
3-Chloro-2-(2-propoxyethoxy)aniline

Hazard: Irritant

Formula: C₁₁H₁₆ClNO₂
MDL Number: MFCD08687471
CAS Number: 946714-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR