N-[2-(2-Amino-6-chlorophenoxy)ethyl]-N-methyl-N-phenylamine

Supplier: Matrix Scientific

021889-500MG
101812-940EA 235.75 USD
101812-940
N-[2-(2-Amino-6-chlorophenoxy)ethyl]-N-methyl-N-phenylamine
N-[2-(2-Amino-6-chlorophenoxy)ethyl]-N-methyl-N-phenylamine

Hazard: Irritant

Formula: C₁₅H₁₇ClN₂O
MDL Number: MFCD08686782
CAS Number: 946772-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR