3-Chloro-2-[2-(4-morpholinyl)ethoxy]phenylamine

Supplier: Matrix Scientific

021886-500MG
101812-934EA 235.75 USD
101812-934
3-Chloro-2-[2-(4-morpholinyl)ethoxy]phenylamine
3-Chloro-2-[2-(4-morpholinyl)ethoxy]phenylamine

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂
MDL Number: MFCD08687399
CAS Number: 946727-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR