2-(Benzyloxy)-3-chloroaniline

Supplier: Matrix Scientific

021883-500MG
101812-928EA 315.38 USD
101812-928
2-(Benzyloxy)-3-chloroaniline
2-(Benzyloxy)-3-chloroaniline

Hazard: Irritant

Formula: C₁₃H₁₂ClNO
MDL Number: MFCD08686861
CAS Number: 59360-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR