3-Chloro-2-ethoxyaniline hydrochloride

Supplier: Matrix Scientific

021880-500MG
101812-922EA 235.75 USD
101812-922
3-Chloro-2-ethoxyaniline hydrochloride
3-Chloro-2-ethoxyaniline hydrochloride

Hazard: Irritant

Formula: C₈H₁₁Cl₂NO
MDL Number: MFCD12405026
CAS Number: 53689-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR